missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyandiamide, 99.5%
CAS: 461-58-5 | C2H4N4 | 84.08 g/mol
Supplier: Thermo Scientific Chemicals 113810025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dicyandiamide | |
| 208.0°C to 211.0°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00008066 | |
| 01,229 | |
| dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin | |
| QGBSISYHAICWAH-UHFFFAOYSA-N | |
| 2-cyanoguanidine | |
| 10005 | |
| 99.5% |
| 461-58-5 | |
| White | |
| 99.5% | |
| C2H4N4 | |
| 2.5 kg | |
| 15, 3103 | |
| Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C) | |
| NC(N)=NC#N | |
| 84.08 | |
| 84.08 | |
| Crystalline Powder |
Chemical Identifiers
| 461-58-5 | |
| 84.08 | |
| QGBSISYHAICWAH-UHFFFAOYSA-N | |
| 10005 |
| C2H4N4 | |
| MFCD00008066 | |
| dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin | |
| NC(N)=NC#N |
Safety and Handling
EINECSNumber : 207-312-8
RUO – Research Use Only