missing translation for 'onlineSavingsMsg'
Learn More

Dicyandiamide, 99.5%

Catalog No. p-3736219
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113810050 5 g Glass Bottle
AC113810250 25 g Plastic bottle
AC113810010 1 kg Plastic bottle
AC113810025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC113810050 Supplier Thermo Scientific Chemicals Supplier No. 113810050
Only null left

CAS: 461-58-5 | C2H4N4 | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
SMILES NC(N)=NC#N

Specifications

Melting Point 208.0°C to 211.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Glass Bottle
Quantity 5 g
Fieser 01,229
Merck Index 15, 3103
Solubility Information Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C)
IUPAC Name 2-cyanoguanidine
Formula Weight 84.08
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material Dicyandiamide
Show More Show Less
Hazard Category
EINECSNumber 207-312-8

RUO – Research Use Only