missing translation for 'onlineSavingsMsg'
Learn More

Dicumarol

Catalog No. p-7042749
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6363406 5 g
AAJ6363414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6363406 Supplier Thermo Scientific Chemicals Supplier No. J6363406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vitamin K reductase inhibitor possessing anticoagulant properties | CAS: 66-76-2 | C19H12O6 | 336.30 g/mol

Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.

Solubility
Soluble in aqueous alkaline solutions, organic bases, 0.1 N NaOH (15 mg/ml), Pyridine (50 mg/ml), chloroform (slightly soluble), and benzene (slightly soluble). Insoluble in water, and alcohols.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 66-76-2
Molecular Formula C19H12O6
Molecular Weight (g/mol) 336.30
MDL Number MFCD00006857
InChI Key DOBMPNYZJYQDGZ-UHFFFAOYSA-N
Synonym dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
PubChem CID 54676038
ChEBI CHEBI:4513
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
SMILES OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12

Specifications

Melting Point 286°C to 289°C
Color White
Quantity 5 g
UN Number UN2811
Merck Index 14,3090
Solubility Information Soluble in aqueous alkaline solutions,organic bases,0.1 N NaOH (15mg/ml),Pyridine (50mg/ml),chloroform (slightly soluble),and benzene (slightly soluble). Insoluble in water,and alcohols.
Formula Weight 336.3
Physical Form Powder
Chemical Name or Material Dicumarol
Hazard Category H301-H372
Hazard Statement GHS H Statement
H372-H302
Causes damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P301+P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-632-9
RTECSNumber GN7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only