missing translation for 'onlineSavingsMsg'
Learn More

Dicumarol, 99%

Catalog No. AC204120050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204120050 5 g Glass bottle
AC204120250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC204120050 Supplier Thermo Scientific Chemicals Supplier No. 204120050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66-76-2 | C19H12O6 | 336.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66-76-2
Molecular Formula C19H12O6
Molecular Weight (g/mol) 336.30
MDL Number MFCD00006857
InChI Key DOBMPNYZJYQDGZ-UHFFFAOYSA-N
Synonym dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
PubChem CID 54676038
ChEBI CHEBI:4513
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
SMILES OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12

Specifications

Melting Point 290.0°C to 292.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 19, 197
Merck Index 15, 3100
Solubility Information Solubility in water: negligible. Other solubilities: practically insoluble in alcohols,ether,aceton,soluble in aq.alkaline solutions,pyridine and,similar org.bases,slightly soluble in benzene and chloroform
Formula Weight 336.29
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Dicumarol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately cal
EINECSNumber 200-632-9
RTECSNumber GN7875000
TSCA TSCA

RUO – Research Use Only