missing translation for 'onlineSavingsMsg'
Learn More

Dicoumarol 98.0+%, TCI America™

Catalog No. p-7132061
Change view
Click to view available options
Quantity:
1 g
25 g
Catalog No. Quantity
M02161G 1 g
M021625G 25 g
2 options

Catalog No. M02161G

Supplier: TCI America M02161G

Only null left

Chemical Identifiers

CAS 66-76-2
Molecular Formula C19H12O6
Molecular Weight (g/mol) 336.30
MDL Number MFCD00006857
InChI Key DOBMPNYZJYQDGZ-UHFFFAOYSA-N
Synonym dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
PubChem CID 54676038
ChEBI CHEBI:4513
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxo-2H-chromen-3-yl)methyl]-2H-chromen-2-one
SMILES OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12

Specifications

Melting Point 288°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 336.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dicoumarol
RTECSNumber GN7875000
TSCA Yes