missing translation for 'onlineSavingsMsg'
Learn More

Dicloxacillin Sodium, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022932
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022932 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022932 Supplier Merck Emd Millipore Supplier No. BP810

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 13412-64-1
Molecular Formula C19H16Cl2N3NaO5S · H2O
Linear Formula C19H16Cl2N3NaO5S · H2O
MDL Number MFCD00167147
Synonym Dicloxacillin sodium salt monohydrate
Type Pharmacopeia and Metrological Institutes Standards
Quantity 250 mg
Formula Weight 510.32
Grade British Pharmacopoeia (BP) Reference Standard