missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac sodium salt

Catalog No. p-7042462
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6260906 5 g
AAJ6260914 25 g
AAJ6260922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6260906 Supplier Thermo Scientific Chemicals Supplier No. J6260906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standard NSAID and cyclooxygenase (COX) inhibitor | CAS: 15307-79-6 | C14H10Cl2NNaO2 | 318.129 g/mol

Oxidation of diclofenac sodium produces the metabolite 4'-hydroxy diclofenac) which demonstrates specific inhibition of Cox-2. Inhibition of Cox by diclofenac and 4'-hydroxy diclofenac suppresses prostaglandin E2 synthesis, producing anti-inflammatory and analgesic effects. Diclofenac is also shown to stabilize the native tetrameric conformation of transthyretin (TTR) fibrils, preventing the formation of insoluble amyloidogenic TTR deposits. Diclofenac Sodium is a substrate of CYP2C9. It is also used as an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxidation of diclofenac sodium produces the metabolite 4′-hydroxy diclofenac) which demonstrates specific inhibition of Cox-2. Inhibition of Cox by diclofenac and 4′-hydroxy diclofenac suppresses prostaglandin E2 synthesis, producing anti-inflammatory and analgesic effects. Diclofenac is also shown to stabilize the native tetrameric conformation of transthyretin (TTR) fibrils, preventing the formation of insoluble amyloidogenic TTR deposits. Diclofenac Sodium is a substrate of CYP2C9. It is also used as an inhibitor of Cox-1 and Cox-2.

Solubility
Soluble in water (50 mg/ml), PBS pH 7.2 (6 mg/ml), ethanol (∼35 mg/ml), DMF (∼35 mg/ml), DMSO (∼35 mg/ml), methanol (>24 mg/ml), and acetone (6 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 15307-79-6
Molecular Formula C14H10Cl2NNaO2
Molecular Weight (g/mol) 318.129
MDL Number MFCD00082251
InChI Key KPHWPUGNDIVLNH-UHFFFAOYSA-M
Synonym diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon
PubChem CID 5018304
ChEBI CHEBI:4509
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+]

Specifications

Melting Point 283°C to 285°C
Color White
Quantity 5 g
UN Number UN2811
Merck Index 14,3081
Solubility Information Soluble in water to 50mg/ml.
Formula Weight 318.14
Physical Form Powder
Chemical Name or Material Diclofenac sodium salt
Hazard Category H301
Hazard Statement GHS H Statement
H301-H361
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 239-346-4
RTECSNumber AG6330000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only