accessibility menu, dialog, popup

UserName

CAS RN 15307-79-6

IUPAC Name: sodium 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
Synonyms: sodium diclofenac
Molecular Weight (g/mol): 318.13
Molecular Formula: C14H10Cl2NNaO2
InChi Key: KPHWPUGNDIVLNH-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)