missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclofenac Sodium Salt 98.0+%, TCI America™
Supplier: TCI America D250825G
Specifications
| Diclofenac Sodium Salt | |
| White-Yellow | |
| MFCD00082251 | |
| 2811 | |
| KPHWPUGNDIVLNH-UHFFFAOYSA-M | |
| sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate | |
| 5018304 | |
| 318.13 | |
| Crystalline Powder |
| 15307-79-6 | |
| C14H10Cl2NNaO2 | |
| 25 g | |
| diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon | |
| C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+] | |
| 318.129 | |
| CHEBI:4509 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 15307-79-6 | |
| 318.129 | |
| KPHWPUGNDIVLNH-UHFFFAOYSA-M | |
| 5018304 | |
| sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate |
| C14H10Cl2NNaO2 | |
| MFCD00082251 | |
| diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon | |
| CHEBI:4509 | |
| C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+] |
Safety and Handling
RTECSNumber : AG6330000
TSCA : No