Learn More
Dickson™ Temperature Recorder Charts
Supplier:  Dickson™ C039
 View more versions of this product
Catalog No. 13940988
Only null left
Description
Description
Specifications
Chemical Identifiers
Temperature Recorder Chart
For the accuracy of data, great way to archive monitoring history
Circular
Specifications
Temperature Recorder Chart
4 in.
Temperature Chart Recorder
0°F to 250°F
7 Days
101 mm
For the accuracy of data, great way to archive monitoring history
Circular
-17.7°C to +121°C
Product Suggestions
Product Suggestions