missing translation for 'onlineSavingsMsg'
Learn More

Dickson™ Datalogger Software

Catalog No. 1390475
Change view
Click to view available options
Accuracy (Temperature):
±1.8°F
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Accuracy (Temperature)
1390475 ±1.8°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1390475 Supplier Dickson™ Supplier No. A015

Includes

Serial download cable (9 pin female D-shell to male 2.5mm stereo plug)

Requires

Microsoft™ Windows 98™ or newer, 4 MB RAM, 1 free COM (serial) port, CD drive

Latest version of DicksonWare™ Software

Use the software to setup your Dickson loggers, view and download data, and reset your logger to the factory settings.

  • Easy installation
  • Easy logger setup and data downloads
  • Data visualization with tables and graphs
  • Real-time monitoring
  • RS-232 COM (serial) port computer interface

Specifications

Type Software and Cable, Dicksonware
Includes Serial download cable
Model A015
Accuracy (Temperature) ±1.8°F