missing translation for 'onlineSavingsMsg'
Learn More

Dicinnamalacetone 98.0+%, TCI America™

Catalog No. D090010G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D090010G 10 g
D090025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D090010G Supplier TCI America Supplier No. D090010G

Chemical Identifiers

CAS 622-21-9
Molecular Formula C21H18O
Molecular Weight (g/mol) 286.374
MDL Number MFCD00004792
InChI Key RLJALOQFYHCJKG-XCBHXTLASA-N
Synonym acetone, dicinnamylidene, 1z,3e,6e,8e-1,9-diphenylnona-1,3,6,8-tetraen-5-one, 1,9-diphenylnona-1,3,6,8-tetraen-5-one, 1,3,6,8-nonatetraen-5-one, 1,9-diphenyl, 1,6,8-nonatetraen-5-one, 1,9-diphenyl, 1z,3e,6e,8e-1,9-diphenyl-5-nona-1,3,6,8-tetraenone
PubChem CID 6436875
IUPAC Name (1Z,3E,6E,8E)-1,9-diphenylnona-1,3,6,8-tetraen-5-one
SMILES C1=CC=C(C=C1)C=CC=CC(=O)C=CC=CC2=CC=CC=C2

Specifications

Melting Point 143°C
Color Yellow
Quantity 10 g
Formula Weight 286.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dicinnamalacetone