missing translation for 'onlineSavingsMsg'
Learn More

μ-Dichlorotetraethylene dirhodium(I)

Catalog No. p-105769
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197591000 100 mg Glass bottle
AC197590010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197591000 Supplier Thermo Scientific Chemicals Supplier No. 197591000
Only null left

CAS: 12081-16-2 | C8H16Cl2Rh2 | 388.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
Molecular Weight (g/mol) 388.93
MDL Number MFCD00013206
InChI Key OOQJCPOXJFCGCR-UHFFFAOYSA-L
Synonym chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i
PubChem CID 45357812
IUPAC Name ethene;rhodium;dichloride
SMILES Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C

Specifications

Color Rust
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble
Formula Weight 388.92
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material µ-Dichlorotetraethylene dirhodium(I)
Hazard Category AIR SENSITIVE
EINECSNumber 235-145-0

RUO – Research Use Only