accessibility menu, dialog, popup

UserName

CAS RN 12081-16-2

IUPAC Name: bis(chlororhodium); tetrakis(ethene)
Synonyms: bis(chlororhodium); tetrakis(ethylene)
Molecular Weight (g/mol): 388.93
Molecular Formula: C8H16Cl2Rh2
InChi Key: OOQJCPOXJFCGCR-UHFFFAOYSA-L
SMILES: Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)