missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylphosphine 98.0+%, TCI America™

Supplier:  TCI America P0207500ML

 View more versions of this product

Catalog No. P0207500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dichlorophenylphosphine
Yellow
C6H5Cl2P
500 mL
dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
ClP(Cl)C1=CC=CC=C1
178.98
178.98
Liquid
644-97-3
59°C
MFCD00000528
2798
IMDXZWRLUZPMDH-UHFFFAOYSA-N
dichloro(phenyl)phosphane
12573
≥98.0% (T)

Chemical Identifiers

644-97-3
178.98
IMDXZWRLUZPMDH-UHFFFAOYSA-N
12573
ClP(Cl)C1=CC=CC=C1
C6H5Cl2P
MFCD00000528
dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
dichloro(phenyl)phosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2517

RTECSNumber : TB2478000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions