missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorophenylphosphine 98.0+%, TCI America™
$40.43 - $273.38
Chemical Identifiers
| CAS | 644-97-3 |
|---|---|
| Molecular Formula | C6H5Cl2P |
| Molecular Weight (g/mol) | 178.98 |
| MDL Number | MFCD00000528 |
| InChI Key | IMDXZWRLUZPMDH-UHFFFAOYSA-N |
| Synonym | dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl |
| PubChem CID | 12573 |
| IUPAC Name | dichloro(phenyl)phosphane |
| SMILES | ClP(Cl)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P020725ML
|
TCI America
P020725ML |
25 mL |
Each for $40.43
|
|
|||||
|
P0207500ML
|
TCI America
P0207500ML |
500 mL |
Each for $273.38
|
|
|||||
Chemical Identifiers
| 644-97-3 | |
| 178.98 | |
| IMDXZWRLUZPMDH-UHFFFAOYSA-N | |
| 12573 | |
| ClP(Cl)C1=CC=CC=C1 |
| C6H5Cl2P | |
| MFCD00000528 | |
| dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl | |
| dichloro(phenyl)phosphane |
Specifications
| 644-97-3 | |
| 59°C | |
| MFCD00000528 | |
| 2798 | |
| IMDXZWRLUZPMDH-UHFFFAOYSA-N | |
| dichloro(phenyl)phosphane | |
| 12573 | |
| ≥98.0% (T) | |
| Dichlorophenylphosphine |
| Yellow | |
| C6H5Cl2P | |
| 25 mL | |
| dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl | |
| ClP(Cl)C1=CC=CC=C1 | |
| 178.98 | |
| 178.98 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2517
RTECSNumber : TB2478000
TSCA : Yes