missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethylvinylsilane 97.0+%, TCI America™
$48.13 - $354.23
Chemical Identifiers
| CAS | 124-70-9 |
|---|---|
| Molecular Formula | C3H6Cl2Si |
| Molecular Weight (g/mol) | 141.066 |
| MDL Number | MFCD00000492 |
| InChI Key | YLJJAVFOBDSYAN-UHFFFAOYSA-N |
| Synonym | dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane |
| PubChem CID | 31299 |
| IUPAC Name | dichloro-ethenyl-methylsilane |
| SMILES | C[Si](C=C)(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D037825ML
|
TCI America
D037825ML |
25 mL |
Each for $48.13
|
|
|||||
|
D0378100ML
|
TCI America
D0378100ML |
100 mL |
Each for $113.59
|
|
|||||
|
D0378500ML
|
TCI America
D0378500ML |
500 mL |
Each for $354.23
|
|
|||||
Chemical Identifiers
| 124-70-9 | |
| 141.066 | |
| YLJJAVFOBDSYAN-UHFFFAOYSA-N | |
| 31299 | |
| C[Si](C=C)(Cl)Cl |
| C3H6Cl2Si | |
| MFCD00000492 | |
| dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane | |
| dichloro-ethenyl-methylsilane |
Specifications
| 124-70-9 | |
| 94°C | |
| MFCD00000492 | |
| 2985 | |
| YLJJAVFOBDSYAN-UHFFFAOYSA-N | |
| dichloro-ethenyl-methylsilane | |
| 31299 | |
| ≥97.0% (GC) | |
| Dichloromethylvinylsilane |
| Yellow | |
| C3H6Cl2Si | |
| 25 mL | |
| dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane | |
| C[Si](C=C)(Cl)Cl | |
| 141.066 | |
| 141.07 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2042
RTECSNumber : VV3533000
TSCA : Yes