accessibility menu, dialog, popup

UserName

CH3SiClClH2C

CAS RN 124-70-9

IUPAC Name: dichloro(ethenyl)methylsilane
Synonyms: silane, dichloroethenylmethyl-
Molecular Weight (g/mol): 141.07
Molecular Formula: C3H6Cl2Si
InChi Key: YLJJAVFOBDSYAN-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)C=C
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)