missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylvinylsilane 97.0+%, TCI America™

Catalog No. D0378500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0378500ML 500 mL
D037825ML 25 mL
D0378100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0378500ML Supplier TCI America Supplier No. D0378500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-70-9
Molecular Formula C3H6Cl2Si
Molecular Weight (g/mol) 141.066
MDL Number MFCD00000492
InChI Key YLJJAVFOBDSYAN-UHFFFAOYSA-N
Synonym dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane
PubChem CID 31299
IUPAC Name dichloro-ethenyl-methylsilane
SMILES C[Si](C=C)(Cl)Cl

Specifications

Color Yellow
Boiling Point 94°C
Quantity 500 mL
UN Number 2985
Formula Weight 141.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloromethylvinylsilane
EINECSNumber (2)-2042
RTECSNumber VV3533000
TSCA Yes