Learn More
Dichloromethylvinylsilane, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 124-70-9 |
|---|---|
| Molecular Formula | C3H6Cl2Si |
| Molecular Weight (g/mol) | 141.07 |
| MDL Number | MFCD00000492 |
| InChI Key | YLJJAVFOBDSYAN-UHFFFAOYSA-N |
| Synonym | dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane |
| PubChem CID | 31299 |
| IUPAC Name | dichloro-ethenyl-methylsilane |
| SMILES | C[Si](C=C)(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC147435000
|
Thermo Scientific Chemicals
147435000 |
500g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 124-70-9 | |
| 141.07 | |
| YLJJAVFOBDSYAN-UHFFFAOYSA-N | |
| 31299 | |
| C[Si](C=C)(Cl)Cl |
| C3H6Cl2Si | |
| MFCD00000492 | |
| dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane | |
| dichloro-ethenyl-methylsilane |
Specifications
| 124-70-9 | |
| 92.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4290 to 1.4310 | |
| MFCD00000492 | |
| 1.08 | |
| Solubility in water: reacts. | |
| C[Si](C=C)(Cl)Cl | |
| 141.07 | |
| 141.07 | |
| Dichloromethylvinylsilane, 97% |
| 1.0800g/mL | |
| 4°C | |
| 96% min. (GC) | |
| C3H6Cl2Si | |
| H2C=CHSi(CH3)Cl2 | |
| 500g | |
| dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane | |
| YLJJAVFOBDSYAN-UHFFFAOYSA-N | |
| dichloro-ethenyl-methylsilane | |
| 31299 | |
| 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 204-710-3
RTECSNumber : VV3533000
TSCA : TSCA
RUO â Research Use Only