missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Stabilized, OmniSolv™, 99.9%, MilliporeSigma™

Catalog No. MDX08311
Change view
Click to view available options
Quantity:
4 Liters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX08311 4 Liters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX08311 Supplier Technidata Supplier No. US1DX0831/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Density 1.32
Quantity 4 Liters
Formula Weight 84.93
Chemical Name or Material Dichloromethane, Stabilized, 99.9%