missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99.9% (GC), Solstice

Catalog No. 60046099
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
4 x 2.5 L
6 x 1 L
250 kg
Packaging:
Glass Bottle
Metal Drum
Stainless Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046099 6 x 1 L Glass Bottle
60046093 1 L Glass Bottle
60046094 2.5 L Glass Bottle
60046098 4 x 2.5 L Glass Bottle
60046091 10 L Stainless Steel Can
60046095 250 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60046099 Supplier Solstice Supplier No. 322226X1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. ISO,≥99.9% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Appearance Liquid
Density 1.325g/mL at 25°C, 1.320 to 1.330g/Lat 20°C
Quantity 6 x 1 L
Explosion Limit 0.22
Ignition Point 1223°F
Packaging Glass Bottle
Refractive Index n20/D 1.4235 to 1.4245
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid: ≤0.001% (as HCl)
Vapor Pressure 6.83psi (20°C), 24.45psi (55°C)
UN Number UN1593
Beilstein 1730800
Boiling Point 39.8°C to 40°C
Formula Weight 84.93g/mol
Percent Purity ≥99.9% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Dichloromethane, Reag. ISO
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
Precautionary Statement H315-H319-H335-H336-H351-H373
DOTInformation 6.1/PGIII
EINECSNumber 200-838-9
RTECSNumber PA8050000