missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Puriss., meets analytical spec. of Ph. Eur., NF, ≥99% (GC), Honeywell Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -97°C |
| Color | Colorless |
| Density | 1.318 to 1.322g/mL at 25°C, 1.325g/mL at 25°C |
| Quantity | 5 L |
| Explosion Limit | 0.22 |
| Ignition Point | 1223°F |
| Packaging | Aluminum Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 24.45psi (55°C), 6.83psi (20°C) |
| UN Number | UN1593 |
| Show More |
Safety and Handling
| Hazard Statement | P261-P281-P305 + P351 + P338 |
| Precautionary Statement | H315-H319-H335-H336-H351-H373 |
| DOTInformation | 6.1/PGIII |
| EINECSNumber | 200-838-9 |
| RTECSNumber | PA8050000 |