missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane/Methanol, 1:1 v/v, Reagents

Catalog No. 01335420
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335420 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335420 Supplier Labchem Inc Supplier No. CD2606004C
Only null left

Dichloromethane/Methanol, 1:1 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-09-2 , 67-56-1
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless to Light Yellow
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.059
Grade Reagent
Chemical Name or Material Dichloromethane/Methanol, 1:1 v/v