missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane (HR-GC), OmniSolv, MilliporeSigma™
Supplier: Technidata DX08371
Specifications
| ™OmniSolv™ Dichloromethane | |
| 75-09-2 | |
| 4 L | |
| CH2Cl2 | |
| MFCD00000881 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93 |
| HR-GC, OmniSolv™ | |
| 1.32 | |
| Liquid | |
| CH2Cl2 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| HR-GC |
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Safety and Handling
Recommended Storage : Room Temperature