missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dichloromethane, HPLC Grade 2L

Catalog No. 402152
Encompass
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
402152 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 402152 Supplier Applied Biosystems™ Supplier No. 402152

This HPLC Grade Dichloromethane comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.

Specifications

Chemical Name or Material Dichloromethane
Content And Storage Store in a cool, dry, and well-ventilated location.
Format Bottle
Sample Type DNA
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
Product Type Dichloromethane Reagent
Quantity 2 L
Purity HPLC
Reagent Type Solvent or General Reagent
Shipping Condition Room Temperature
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.