missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, GC2™ , for GC, pesticide residue analysis, contains amylene, >99.9%, Honeywell Burdick & Jackson
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 1.33 g/cm3 |
| Quantity | 200 L |
| Packaging | Returnable Steel Drum |
| UN Number | UN1593 |
| Beilstein | 1730800 |
| Boiling Point | 40°C |
| Formula Weight | 84.94 |
| Percent Purity | >99.9% |
| Grade | GC/Pesticide Residue Analysis |
| Show More |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane |