missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, For pesticide residue analysis, Solstice

Catalog No. 6000499
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000499 1 L
6000500 2.5 L
60046348 4 x 2.5 L
6000502 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000499 Supplier Solstice Supplier No. 344881L

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Density 1.325g/mL at 25°C
Quantity 1 L
Explosion Limit 0.22
Ignition Point 1223°F
Vapor Pressure 24.45 psi (55°C), 6.83 psi (20°C)
UN Number UN1593
Beilstein 1730800
Formula Weight 84.93g/mol
Percent Purity ≥99.8% (GC)
Grade CHROMASOLV
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Statement P260-P280-P305 + P351 + P338
RTECSNumber PA8050000