missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane For gas chromatography MS SupraSolv™, MilliporeSigma™

Catalog No. M1006682500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1006682500 2.5 L Glass Bottle
M1006681000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1006682500 Supplier Technidata Supplier No. 1.00668.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography MS SupraSolv™

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

pH Neutral (at 20°C)
Melting Point -95°C
Color Colorless
Density 1.33g/cm3 at 20°C
Quantity 2.5 L
Ignition Point 605°C (1121°F)
Odor Sweet
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.42
Vapor Pressure 475 hPa (20°C)
UN Number 1593
Boiling Point 40°C
Solubility Information Soluble in water: 20g/L at 20°C
Formula Weight 84.93
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category H351: Suspected of causing cancer.
Recommended Storage 5°C to 30°C (41°F to 86°F), Tightly closed. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons. Protected from light.