missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, MilliporeSigma™
$164.48 - $345.04
Chemical Identifiers
| CAS | 75-09-2 |
|---|---|
| Molecular Formula | CH2Cl2 |
| Molecular Weight (g/mol) | 84.93 |
| MDL Number | MFCD00000881 |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| IUPAC Name | dichloromethane |
| SMILES | ClCCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
MDX08316
|
Technidata
DX08316 |
1 L | Glass Bottle |
Each for $164.48
|
|
||||
|
MDX08381
|
Technidata
DX08381 |
4 L | Glass Bottle |
Each for $283.52
|
|
||||
|
MDX0838P1
|
Technidata
DX0838P1 |
4 L | Poly-coated Glass Bottle |
Each for $345.04
|
|
||||
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Specifications
| 75-09-2 | |
| Glass Bottle | |
| CH2Cl2 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| Dichloromethane |
| 1 L | |
| CH2Cl2 | |
| MFCD00000881 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93 |