missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, MilliporeSigma™
Specifications
Specifications
| Packaging | Poly-coated Glass Bottle |
| Linear Formula | CH2Cl2 |
| Formula Weight | 84.93 |
| Chemical Name or Material | Dichloromethane |