missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane CHROMASOLV™, for HPLC, ≥99.8%, contains amylene as stabilizer, Solstice

Catalog No. 6000495
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000495 4 x 2.5 L
6000492 1 L
6000493 2.5 L
6000497 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000495 Supplier Solstice Supplier No. 348564X2.5L
Only null left

Product filtered through a 0.2μm filter

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Density 1.325g/mL at 25°C
Quantity 4 x 2.5 L
Refractive Index n20/D 1.424
Linear Formula CH2Cl2
Vapor Pressure 6.83 psi (20°C)
Beilstein 1730800
Boiling Point 39.8°C to 40°C
Formula Weight 84.93g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material Dichloromethane, for HPLC, contains amylene as stabilizer
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
Precautionary Statement H315-H319-H335-H336-H351-H373
EINECSNumber 200-838-9
RTECSNumber PA8050000