missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane CHROMASOLV™, for HPLC, ≥99.8%, contains amylene as stabilizer, Solstice
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -97°C |
| Density | 1.325g/mL at 25°C |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.424 |
| Linear Formula | CH2Cl2 |
| Vapor Pressure | 6.83 psi (20°C) |
| Beilstein | 1730800 |
| Boiling Point | 39.8°C to 40°C |
| Formula Weight | 84.93g/mol |
| Percent Purity | ≥99.8% |
| Show More |
Safety and Handling
| Hazard Statement | P261-P281-P305 + P351 + P338 |
| Precautionary Statement | H315-H319-H335-H336-H351-H373 |
| EINECSNumber | 200-838-9 |
| RTECSNumber | PA8050000 |