missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, B&J Brand™, for HPLC, spectrophotometry (not for pesticide residue analysis), contains cyclohexene , >99.9%, Honeywell Burdick & Jackson

$7667.65 - $7667.65

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6000504
View Documents
Honeywell Chemicals
CS300200
200 L Returnable Steel Drum
Each for $7,667.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

75-09-2
84.93
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
CHEBI:15767
ClCCl

Specifications

75-09-2
100.0000%
40°C
Returnable Steel Drum
MFCD00000881
0.005% max.
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.94
HPLC
0.1 AU max. at 240 nm, 0.01 AU max. at 250 nm
1.33 g/cm3
200 L
CH2Cl2
UN1593
1730800
160% in Water at 20°C
ClCCl
84.93
CHEBI:15767
>99.9%
Dichloromethane, B&J Brand™, for HPLC, spectrophotometry (not for pesticide residue analysis), contains cyclohexene , >99.9%
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane

SDS
missing translation for 'documents'

missing translation for 'documents'