missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains cyclohexene , >99.9%, Honeywell Burdick & Jackson

Catalog No. 6000510
Change view
Click to view available options
Quantity:
4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000510 6 x 1 L Glass Bottle
6000507 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000510 Supplier Honeywell Chemicals Supplier No. 3001L
Only null left

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

CAS Max % 100.0000%
Density 1.33 g/cm3
Quantity 6 x 1 L
Packaging Glass Bottle
UN Number UN1593
Beilstein 1730800
Boiling Point 40°C
Formula Weight 84.94
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains cyclohexene , >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane