missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains amylene, >99.9%, Honeywell Burdick & Jackson
Supplier: Honeywell Chemicals CS299200
Specifications
| Dichloromethane | |
| 75-09-2 | |
| 100.0000% | |
| 40°C | |
| Returnable Steel Drum | |
| MFCD00000881 | |
| 0.01% max. | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.94 | |
| HPLC |
| B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains amylene, >99.9% | |
| 0.1 AU max. at 240 nm, 0.01 AU max. at 250 nm | |
| 1.33 g/cm3 | |
| 200 L | |
| CH2Cl2 | |
| UN1593 | |
| 1730800 | |
| 160% in Water at 20°C | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| >99.9% |
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane