missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane Anhydrous (stabilized with 2-Methyl-2-butene) 99.0+%, TCI America™

Catalog No. D3478500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3478500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3478500ML Supplier TCI America Supplier No. D3478500ML
Only null left

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Boiling Point 40°C
Quantity 500 mL
UN Number 1593
Formula Weight 84.93
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloromethane Anhydrous (stabilized with 2-Methyl-2-butene)
EINECSNumber (2)-0036
RTECSNumber PA8050000
TSCA Yes