missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Anhydrous, 99.8% min., DriSolv, MilliporeSigma™

Catalog No. p-4382176
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX08347 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX08347 Supplier Technidata Supplier No. DX08347
Only null left

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Quantity 100 mL
Physical Form Liquid
Assay Percent Range 99.8%, min.
Linear Formula K2HPO4
Formula Weight 174.18
Chemical Name or Material Dichloromethane, Anhydrous, DriSolv™