missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene

Catalog No. AA41835AK
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41835AK 250 mL
AA41835K2 1 L
AA41835M1 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41835AK Supplier Thermo Scientific Chemicals Supplier No. 041835AK

Solvent For Anhydrous Reactions, Water <0.01% | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

Solvent for anhydrous reactions, water <0.01%.Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point - 95°C
Color Colorless
Density 1.325 g/cm3 at 20°C
Boiling Point 39°C to 40°C
Ignition Point 605°C (1121°F)
Odor Sweet
Assay Percent Range >99.7%
Refractive Index 1.4244
Quantity 250 mL
Beilstein 1730800
Solubility Information Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water.
Vapor Pressure 475 hPa (356mm Hg) at 20°C
Formula Weight 84.93
Physical Form Liquid
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category H315-H319-H336-H350-H373
Precautionary Statement P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 200-838-9
RTECSNumber PA8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only