missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS Reagent (Methylene Chloride), Reagents

Catalog No. p-200021096
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338020 1 L
01338019 2.5 L
01338018 200 L
01338017 20 L
01338016 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01338020 Supplier Labchem Inc Supplier No. C11959001C
Only null left

Dichloromethane, ACS Reagent (Methylene Chloride), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Methylene Chloride
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -96.7°C
Color Colorless
Boiling Point 39.8°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.3266
Grade ACS
Chemical Name or Material Dichloromethane, ACS Reagent (Methylene Chloride)