missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS Reagent (Methylene Chloride), Reagents

Catalog No. 01338020
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338020 1 L
01338019 2.5 L
01338018 200 L
01338017 20 L
01338016 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01338020 Supplier Labchem Inc Supplier No. C11959001C

Dichloromethane, ACS Reagent (Methylene Chloride), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Methylene Chloride
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -96.7°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.3266
Boiling Point 39.8°C
Grade ACS
Chemical Name or Material Dichloromethane, ACS Reagent (Methylene Chloride)