missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCD01001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCD01001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCD01001C Supplier Ricca Chemical Company Supplier No. RDCD01001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methylene Chloride

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

CAS Max % 100
Appearance Colorless Liquid
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.002%
Titratable Acid max. 0.003 meq/g
Water max. 0.02%
Boiling Point 39.8°C
Percent Purity min. 99.5%
Grade ACS
Chemical Name or Material Dichloromethane
Show More Show Less
ShelfLife 36 months