missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS, 99.5+%, stab. with amylene

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, Free halogens P.T., H2O 0.02% | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

$70.26 - $263.03

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
IUPAC Name dichloromethane
SMILES ClCCl
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39116AP
View Documents
Thermo Scientific Chemicals
039116AP
500 mL
Each for $70.26
Only null left
 
AA39116K2
View Documents
Thermo Scientific Chemicals
039116K2
1 L
Each for $102.74
Only null left
 
AA39116K4
View Documents
Thermo Scientific Chemicals
039116K4
2.5 L
Each for $169.11
Only null left
 
AA39116M1
View Documents
Thermo Scientific Chemicals
039116M1
4 x 1 L
Each for $263.03
Only null left
 
AA39116K7
View Documents
Thermo Scientific Chemicals
039116K7
4 L
Each for $243.62
Only null left
 
Description

Description

Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-09-2
84.93
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
CH2Cl2
MFCD00000881
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
ClCCl

Specifications

75-09-2
Colorless liquid
39°C to 40°C
605°C (1121°F)
Liquid
CH2Cl2
475 hPa (356mm Hg) at 20°C
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
84.93
Dichloromethane
Colorless
1.325 g/cm3 at 20°C
500 mL
Sweet
1.4244
MFCD00000881
1730800
Miscible in ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform.
ClCCl
84.93
ACS
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H336-H350-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 200-838-9

RTECSNumber : PA8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only