missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS, 99.5%, Spectrum™ Chemical

Catalog No. p-7165866
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610186 100 mL Amber Glass Bottle
18610188 500 mL Amber Glass Bottle
18610184 4 L Amber Glass Bottle
18610187 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610186 Supplier Spectrum Chemical Mfg Cor Supplier No. M1250100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-09-2
Molecular Weight (g/mol) 84.93

Specifications

CAS Max % 1
Quantity 100 mL
Identification To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Titratable Acid 0.0003 meq/g
Water 0.0002
Boiling Point 39.8°C
Percent Purity 0.995
Grade Reagent
Chemical Name or Material Dichloromethane
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.