missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99+%, extra pure, stabilized with amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
$191.00 - $191.00
Chemical Identifiers
| CAS | 75-09-2 |
|---|---|
| Molecular Formula | CH2Cl2 |
| Molecular Weight (g/mol) | 84.93 |
| MDL Number | MFCD00000881 |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| IUPAC Name | dichloromethane |
| SMILES | ClCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC383780025
|
Thermo Scientific Chemicals
383780025 |
2.5 L |
Each for $191.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Specifications
| -97.0°C | |
| 1.3250g/mL | |
| 2.5 L | |
| 99+% | |
| CH2Cl2 | |
| MFCD00000881 | |
| 01,676; 02,273; 04,337; 07,239; 13,106 | |
| Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| 99+% | |
| Dichloromethane |
| 75-09-2,513-35-9 | |
| 39.0°C to 40.0°C | |
| Glass Bottle | |
| 1.325 | |
| CH2Cl2 | |
| 0.43 mPa.s (20°C) | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93 | |
| Extra Pure |
Safety and Handling
EINECSNumber : 200-838-9
RUO – Research Use Only