Learn More
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Supplier: Thermo Scientific Chemicals 326760025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysisSpecifications
| Dichloromethane | |
| 75-09-2,513-35-9 | |
| 0.001% max. (HCl) | |
| 1.3250g/mL | |
| 2.5 L | |
| 0.0005% max. | |
| 1.325 | |
| CH2Cl2 | |
| 0.43 mPa.s (20°C) | |
| 01,676; 02,273; 04,337; 07,239; 13,106 | |
| Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| 99.9% |
| -97.0°C | |
| 0.5 max. at 235nm, 0.108 max. at 240nm, 0.009 max. at 260nm | |
| Clear liquid | |
| 39.0°C to 40.0°C | |
| Glass Bottle | |
| 99.9% | |
| CH2Cl2 | |
| MFCD00000881 | |
| 0.02% max. (Coulometric) | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93 | |
| Residue Analysis |
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Safety and Handling
GHS H Statement
Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause drowsiness or dizziness.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.
Warning
RUO – Research Use Only