missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.9%, MilliporeSigma™

Catalog No. 50947846
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947846 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947846 Supplier Technidata Supplier No. DX0831P1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point −95°C
Quantity 4 L
Packaging Glass Bottle (Pre-packed)
Residue after Evaporation 1 ppm max
Assay Percent Range 99.9% min
Vapor Pressure 475 hPa (20°C)
Solubility Information 20 g/L
Recommended Storage Store at room temperature