missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019477
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019477 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019477 Supplier Merck Emd Millipore Supplier No. 025755ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dichloromethane (methylene chloride) is a clear, colorless, non-flammable, volatile liquid chlorinated hydrocarbon with a sweet, pleasant smell and emits highly toxic fumes of phosgene when heated to decomposition. It is primarily used as a solvent in paint removers, in aerosol formulations, and as a solvent in the manufacture of pharmaceuticals. It is also used as a degreasing agent in electronics manufacturing and as an ethane foam blowing agent. Above all that it is used as an extraction solvent in the preparation of decaffeinated coffee, hop extracts and spice oleoresins. Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 75-09-2
Ignition Point 1223°F
Solution Contains 0.002% amylene as stabilizer
Vapor Pressure 24.45 psi (55°C)
Vapor Density 2.9 (vs. air)
Density 1.325 g/mL (at 25°C)
Linear Formula CH2 Cl2
MDL Number MFCD00000881
Melting Point -97°C
Boiling Point 39.8°C to 40°C
Synonym Methylene chloride
Type Analytical Standard
Quantity 5 mL
Formula Weight 84.93
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less