missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.5% min., ACS, MilliporeSigma™

Catalog No. MDX08356
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX08356 500 mL
MDX08353 4 L
MDX08355 20 L
MDX083520 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MDX08356 Supplier Technidata Supplier No. DX08356

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 99.5% min.
Formula Weight 84.93
Chemical Name or Material Dichloromethane, For ACS analysis