missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiisopropylsilane 98.0+%, TCI America™

Catalog No. D26035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26035G 5 g
D260325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26035G Supplier TCI America Supplier No. D26035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7751-38-4
Molecular Formula C6H14Cl2Si
Molecular Weight (g/mol) 185.16
MDL Number MFCD00054895
InChI Key GSENNYNYEKCQGA-UHFFFAOYSA-N
Synonym diisopropyldichlorosilane, dichlorodiisopropylsilane, silane, dichlorobis 1-methylethyl, dichloro-di propan-2-yl silane, silane, dichlorodiisopropyl, acmc-209pad, dichloro diisopropyl silane, di 2-propyl dichlorosilane, dichlorobis propan-2-yl silane, bis chloranyl-di propan-2-yl silane
PubChem CID 2758384
IUPAC Name dichlorobis(propan-2-yl)silane
SMILES CC(C)[Si](Cl)(Cl)C(C)C

Specifications

Boiling Point 170°C
Quantity 5 g
UN Number 2985
Formula Weight 185.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorodiisopropylsilane
EINECSNumber (2)-2041
TSCA No